Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108034.1 | NP_001101504.1 | 9 - 10 | 1546 | 156 | |
XM_006240258.2 | XP_006240320.1 | 10 - 11 | 2115 | 156 | ||
XM_008764773.1 | XP_008762995.1 | 10 - 11 | 1719 | 156 | ||
XM_008764774.1 | XP_008762996.1 | 10 - 11 | 1483 | 156 | ||
GenBank mRNA | BC168247.1 | - | 10 - 11 | 1604 | 5597968 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |