Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcaf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100718.1 | NP_001094188.1 | 5 - 6 | 683 | 99 | |
XM_006240279.2 | XP_006240341.1 | 5 - 6 | 485 | 99 | ||
XM_008764782.1 | XP_008763004.1 | 5 - 6 | 538 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |