Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Seli | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134754.1 | NP_001128226.2 | 7 - 8 | 817 | 78 | |
GenBank mRNA | BC168235.1 | - | 7 - 8 | 817 | 7444430 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |