Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dync2li1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013940.1 | NP_001013962.1 | 8 - 9 | 732 | 73 | |
XM_006239695.2 | XP_006239757.1 | 8 - 9 | 787 | 73 | ||
GenBank mRNA | BC079201.1 | - | 8 - 9 | 732 | 7131728 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |