Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbks | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108703.1 | NP_001102173.1 | 3 - 4 | 321 | 98 | |
XM_006239805.2 | XP_006239867.1 | 3 - 4 | 367 | 98 | ||
XM_006239806.2 | XP_006239868.1 | 3 - 4 | 367 | 98 | ||
XM_008764520.1 | XP_008762742.1 | 3 - 4 | 815 | 98 | ||
XM_008764521.1 | XP_008762743.1 | 3 - 4 | 367 | 98 | ||
GenBank mRNA | FQ209797.1 | - | 3 - 4 | 303 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |