Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270619.1 | NP_001257548.1 | 5 - 6 | 1542 | 99 | |
GenBank mRNA | AJ271834.1 | - | 5 - 6 | 1535 | 99 | |
BC061986.1 | - | 5 - 6 | 1497 | 5599216 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Protein Phosphatase 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |