Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fntb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172034.2 | NP_742031.1 | 4 - 5 | 423 | 85 | |
GenBank mRNA | BC087675.1 | - | 4 - 5 | 423 | 7319293 | 85 |
M69056.1 | - | 4 - 5 | 437 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |