Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Preb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170708.1 | NP_001164179.1 | 2 - 3 | 492 | 99 | |
GenBank mRNA | BC078936.1 | - | 2 - 3 | 492 | 7108249 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |