Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spast | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108702.2 | NP_001102172.2 | 12 - 13 | 1432 | 115 | |
XM_008764486.1 | XP_008762708.1 | 13 - 14 | 1808 | 115 | ||
XM_008764487.1 | XP_008762709.1 | 13 - 14 | 1805 | 115 | ||
XM_008764488.1 | XP_008762710.1 | 12 - 13 | 1713 | 115 | ||
GenBank mRNA | BC166846.1 | - | 12 - 13 | 1432 | 9030077 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |