Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcea1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025735.1 | NP_001020906.1 | 4 - 5 | 408 | 107 | |
XM_006237816.2 | XP_006237878.1 | 4 - 5 | 469 | 107 | ||
XM_008763546.1 | XP_008761768.1 | 4 - 5 | 296 | 107 | ||
GenBank mRNA | BC099141.1 | - | 4 - 5 | 408 | 7460791 | 107 |
FQ227501.1 | - | 4 - 5 | 380 | 107 |
Gene Symbol: LOC498453 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251720.2 | XP_006251782.1 | 1 | 462 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |