Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001056014.5 | XP_001056014.2 | 540 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |