Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276434.1 | NP_001263363.1 | 1 - 2 | 194 | 55 | |
GenBank mRNA | BE121399.1 | - | 5 - 6 | 55 | ||
DY318327.1 | - | 1 - 2 | 93 | 55 |
Gene Symbol: Hmgn5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134706.1 | NP_001128178.1 | 1 - 2 | 194 | 55 | |
GenBank mRNA | BC168165.1 | - | 1 - 2 | 96 | 8370295 | 55 |
BE121400.1 | - | 5 - 6 | 55 | |||
BF289419.1 | - | 1 - 2 | 211 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |