Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apool | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014105.1 | NP_001014127.1 | 3 - 4 | 377 | 155 | |
XM_006257191.2 | XP_006257253.1 | 3 - 4 | 316 | 155 | ||
GenBank mRNA | BC086434.1 | - | 3 - 4 | 377 | 7302620 | 155 |
FQ219134.1 | - | 3 - 4 | 267 | 155 | ||
FQ235331.1 | - | 3 - 4 | 263 | 155 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |