Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timmdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007676.1 | NP_001007677.1 | 3 - 4 | 550 | 76 | |
GenBank mRNA | BC079140.1 | - | 3 - 4 | 550 | 7127856 | 76 |
FQ215140.1 | - | 3 - 4 | 577 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |