Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsamp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006248339.2 | XP_006248401.1 | 7 - 8 | 1457 | 84 | |
GenBank mRNA | AY326256.1 | - | 7 - 8 | 975 | 84 | |
BC087607.1 | - | 7 - 8 | 977 | 7314244 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |