Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcls1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011898.1 | NP_001011898.1 | 9 - 10 | 727 | 81 | |
GenBank mRNA | BC079081.1 | - | 9 - 10 | 727 | 7122756 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |