Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upk1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024253.1 | NP_001019424.1 | 4 - 5 | 393 | 67 | |
XM_006248367.2 | XP_006248429.1 | 4 - 5 | 600 | 67 | ||
XM_006248368.2 | XP_006248430.1 | 4 - 5 | 440 | 67 | ||
GenBank mRNA | BC093613.1 | - | 4 - 5 | 393 | 7309649 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |