Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qtrtd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006248320.2 | XP_006248382.1 | 4 - 5 | 521 | 108 | |
XM_008768759.1 | XP_008766981.1 | 4 - 5 | 521 | 108 | ||
XM_008768760.1 | XP_008766982.1 | 4 - 5 | 521 | 108 | ||
XM_008768761.1 | XP_008766983.1 | 4 - 5 | 521 | 108 | ||
XM_008768762.1 | XP_008766984.1 | 2 - 3 | 139 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |