Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fstl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024369.2 | NP_077345.1 | 8 - 9 | 776 | 83 | |
GenBank mRNA | AY325203.1 | - | 1154 | 83 | ||
BC087014.1 | - | 8 - 9 | 776 | 7189359 | 83 | |
U06864.1 | - | 8 - 9 | 751 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |