Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ripk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107103.1 | NP_001100573.1 | 5 - 6 | 875 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Rattus norvegicus Probe spans exons |