Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138882.1 | NP_620237.1 | 6 - 7 | 769 | 75 | |
XM_006248377.2 | XP_006248439.1 | 6 - 7 | 1178 | 75 | ||
GenBank mRNA | BC078727.1 | - | 6 - 7 | 769 | 7124402 | 75 |
D88666.1 | - | 6 - 7 | 769 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |