Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012144.1 | NP_001012144.1 | 5 - 6 | 844 | 132 | |
XM_006248236.2 | XP_006248298.1 | 5 - 6 | 970 | 132 | ||
XM_006248237.2 | XP_006248299.1 | 5 - 6 | 970 | 132 | ||
GenBank mRNA | AB218900.1 | - | 4 - 5 | 585 | 132 | |
BC078947.1 | - | 5 - 6 | 844 | 7109719 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Rattus norvegicus Probe spans exons |