Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap17b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002730183.4 | XP_002730229.1 | 4 - 5 | 1207 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Rattus norvegicus 1700 Probe spans exons |