Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcts1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044237.1 | NP_001037702.1 | 2 - 3 | 717 | 144 | |
XM_006257488.2 | XP_006257550.1 | 2 - 3 | 546 | 144 | ||
GenBank mRNA | BC098848.1 | - | 2 - 3 | 717 | 7379624 | 144 |
FQ211690.1 | - | 1 - 2 | 152 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |