Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsks | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199088.1 | NP_954519.1 | 1 - 2 | 189 | 100 | |
XM_006229014.2 | XP_006229076.1 | 1 - 2 | 251 | 100 | ||
XM_006229015.2 | XP_006229077.1 | 1 - 2 | 251 | 100 | ||
XM_006229017.2 | XP_006229079.1 | 1 - 2 | 250 | 100 | ||
XM_006229018.2 | XP_006229080.1 | 1 - 2 | 250 | 100 | ||
XM_006229019.2 | XP_006229081.1 | 1 - 2 | 250 | 100 | ||
XM_006229020.2 | XP_006229082.1 | 1 - 2 | 250 | 100 | ||
GenBank mRNA | AY346104.1 | - | 1 - 2 | 189 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |