Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004258.1 | NP_001004258.1 | 1 - 2 | 698 | 94 | |
XM_008759161.1 | XP_008757383.1 | 2 - 3 | 1625 | 94 | ||
GenBank mRNA | BC079088.1 | - | 1 - 2 | 698 | 7123358 | 94 |
FQ215228.1 | - | 1 - 2 | 793 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |