Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gltscr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106226.2 | NP_001099696.2 | 5 - 6 | 2293 | 120 | |
XM_006228343.2 | XP_006228405.1 | 7 - 8 | 2640 | 120 | ||
XM_006228344.2 | XP_006228406.1 | 6 - 7 | 2574 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |