Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haus5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002728731.3 | XP_002728777.1 | 13 - 14 | 1241 | 79 | |
XM_008759256.1 | XP_008757478.1 | 13 - 14 | 1233 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |