Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haus5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002728731.3 | XP_002728777.1 | 18 - 19 | 1875 | 54 | |
XM_008759256.1 | XP_008757478.1 | 18 - 19 | 1867 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |