Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyrobp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212525.1 | NP_997690.1 | 2 - 3 | 95 | 128 | |
GenBank mRNA | AY247021.1 | - | 2 - 3 | 95 | 128 | |
FQ211365.1 | - | 2 - 3 | 101 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |