Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207592.1 | NP_997475.1 | 11 - 12 | 982 | 139 | |
GenBank mRNA | BC062005.1 | - | 11 - 12 | 982 | 5599776 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |