Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hrc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181369.2 | NP_852034.1 | 1 - 2 | 2069 | 83 | |
GenBank mRNA | AY277279.1 | - | 1 - 2 | 2069 | 83 | |
FQ216289.1 | - | 1 - 2 | 2163 | 83 | ||
FQ224148.1 | - | 1 - 2 | 2155 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |