Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam98c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198584.1 | NP_001185513.1 | 3 - 4 | 353 | 86 | |
NR_037147.1 | 2 - 3 | 428 | 86 | |||
XM_006228717.2 | XP_006228779.1 | 3 - 4 | 383 | 86 | ||
GenBank mRNA | BC079080.1 | - | 2 - 3 | 391 | 7122702 | 86 |
CO564588.1 | - | 1 - 2 | 130 | 86 | ||
CO570848.1 | - | 3 - 4 | 353 | 86 | ||
DV214726.1 | - | 3 - 4 | 378 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |