Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Izumo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017514.1 | NP_001017514.1 | 8 - 9 | 1052 | 129 | |
XM_008759406.1 | XP_008757628.1 | 7 - 8 | 1194 | 129 | ||
GenBank mRNA | AB195683.1 | - | 7 - 8 | 816 | 129 | |
BC079244.1 | - | 8 - 9 | 1052 | 7132791 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |