Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039665.1 | NP_001034754.1 | 4 - 5 | 727 | 88 | |
XM_006229031.2 | XP_006229093.1 | 4 - 5 | 628 | 88 | ||
XM_006229032.2 | XP_006229094.1 | 4 - 5 | 593 | 88 | ||
XM_006229033.2 | XP_006229095.1 | 4 - 5 | 959 | 88 | ||
XM_008759338.1 | XP_008757560.1 | 4 - 5 | 556 | 88 | ||
GenBank mRNA | AY827147.1 | - | 4 - 5 | 727 | 88 | |
AY827148.1 | - | 4 - 5 | 596 | 88 | ||
BC127438.1 | - | 1401 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |