Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173294.2 | NP_775416.1 | 3 - 4 | 519 | 73 | |
GenBank mRNA | BC088103.1 | - | 3 - 4 | 484 | 73 | |
BC105823.1 | - | 3 - 4 | 484 | 73 | ||
BC127479.1 | - | 3 - 4 | 500 | 7372256 | 73 | |
FQ210362.1 | - | 3 - 4 | 508 | 73 | ||
M20406.1 | - | 3 - 4 | 519 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |