Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Meis3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108472.1 | NP_001101942.1 | 13 - 14 | 1356 | 117 | |
XM_006228346.2 | XP_006228408.1 | 11 - 12 | 1597 | 117 | ||
XM_006228347.2 | XP_006228409.1 | 12 - 13 | 2130 | 117 | ||
XM_006228348.2 | XP_006228410.1 | 11 - 12 | 1570 | 117 | ||
XM_006228349.2 | XP_006228411.1 | 12 - 13 | 1444 | 117 | ||
XM_006228350.2 | XP_006228412.1 | 12 - 13 | 1393 | 117 | ||
XM_006228351.2 | XP_006228413.1 | 12 - 13 | 1366 | 117 | ||
XM_008758950.1 | XP_008757172.1 | 12 - 13 | 1417 | 117 | ||
GenBank mRNA | BC160854.1 | - | 11 - 12 | 1470 | 8364874 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |