Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Meis3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108472.1 | NP_001101942.1 | 4 - 5 | 554 | 144 | |
XM_006228346.2 | XP_006228408.1 | 3 - 4 | 615 | 144 | ||
XM_006228347.2 | XP_006228409.1 | 4 - 5 | 1148 | 144 | ||
XM_006228348.2 | XP_006228410.1 | 3 - 4 | 615 | 144 | ||
XM_006228349.2 | XP_006228411.1 | 3 - 4 | 615 | 144 | ||
XM_006228350.2 | XP_006228412.1 | 3 - 4 | 615 | 144 | ||
XM_006228351.2 | XP_006228413.1 | 3 - 4 | 615 | 144 | ||
XM_008758950.1 | XP_008757172.1 | 3 - 4 | 615 | 144 | ||
GenBank mRNA | BC160854.1 | - | 3 - 4 | 515 | 8364874 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |