Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Meis3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108472.1 | NP_001101942.1 | 9 - 10 | 1068 | 125 | |
XM_006228346.2 | XP_006228408.1 | 7 - 8 | 1309 | 125 | ||
XM_006228347.2 | XP_006228409.1 | 8 - 9 | 1842 | 125 | ||
XM_006228348.2 | XP_006228410.1 | 7 - 8 | 1282 | 125 | ||
XM_006228349.2 | XP_006228411.1 | 8 - 9 | 1156 | 125 | ||
XM_006228350.2 | XP_006228412.1 | 8 - 9 | 1105 | 125 | ||
XM_006228351.2 | XP_006228413.1 | 8 - 9 | 1078 | 125 | ||
XM_008758950.1 | XP_008757172.1 | 8 - 9 | 1129 | 125 | ||
GenBank mRNA | BC160854.1 | - | 7 - 8 | 1182 | 8364874 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |