Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nucb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053463.1 | NP_445915.1 | 5 - 6 | 664 | 62 | |
XM_006229163.1 | XP_006229225.1 | 6 - 7 | 784 | 62 | ||
XM_006229164.2 | XP_006229226.1 | 6 - 7 | 728 | 62 | ||
XM_006229165.1 | XP_006229227.1 | 6 - 7 | 781 | 62 | ||
GenBank mRNA | AF336828.1 | - | 5 - 6 | 664 | 62 | |
BC100643.1 | - | 6 - 7 | 714 | 7930904 | 62 | |
Z36277.1 | - | 6 - 7 | 726 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |