Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017124.1 | NP_058820.1 | 1 - 2 | 133 | 79 | |
XM_006228981.2 | XP_006229043.1 | 2 - 3 | 326 | 79 | ||
XM_008759326.1 | XP_008757548.1 | 1 - 2 | 176 | 79 | ||
GenBank mRNA | BC059144.1 | - | 1 - 2 | 174 | 6887407 | 79 |
FQ230385.1 | - | 1 - 2 | 151 | 79 | ||
X53517.1 | - | 1 - 2 | 133 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |