Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krtdap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB011028.1 | - | 218 | 195 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |