Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012067.1 | NP_001012067.1 | 4 - 5 | 740 | 93 | |
GenBank mRNA | BC083883.1 | - | 4 - 5 | 740 | 7131596 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |