Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pepd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009641.1 | NP_001009641.1 | 8 - 9 | 637 | 76 | |
GenBank mRNA | BC088440.1 | - | 8 - 9 | 637 | 7126171 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |