Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E2f8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229263.2 | XP_006229325.1 | 9 - 10 | 2195 | 86 | |
GenBank mRNA | BC099080.1 | - | 4 - 5 | 544 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |