Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008759356.1 | XP_008757578.1 | 1 - 2 | 84 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |