Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fsd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191627.1 | NP_001178556.1 | 9 - 10 | 1673 | 97 | |
XM_006229440.2 | XP_006229502.1 | 10 - 11 | 1877 | 97 | ||
XM_008759537.1 | XP_008757759.1 | 10 - 11 | 3094 | 97 | ||
XM_008759538.1 | XP_008757760.1 | 11 - 12 | 2267 | 97 | ||
GenBank mRNA | FQ223649.1 | - | 10 - 11 | 1825 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Probe spans exons |