Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017181.2 | NP_058877.1 | 3 - 4 | 421 | 80 | |
GenBank mRNA | BC076381.1 | - | 3 - 4 | 421 | 7108528 | 80 |
FQ219602.1 | - | 3 - 4 | 379 | 80 | ||
M77694.1 | - | 3 - 4 | 343 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |