Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011932.1 | NP_001011932.1 | 1 - 2 | 461 | 98 | |
GenBank mRNA | BC085850.1 | - | 1 - 2 | 461 | 7189751 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |