Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrna7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012832.3 | NP_036964.3 | 4 - 5 | 352 | 70 | |
XM_008759497.1 | XP_008757719.1 | 2 - 3 | 4210 | 70 | ||
GenBank mRNA | AY574256.1 | - | 4 - 5 | 352 | 70 | |
L31619.1 | - | 4 - 5 | 374 | 70 | ||
S53987.1 | - | 393 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Rattus norvegicus Probe spans exons |